Learn More
3-Amino-6-nitro-4-phenyl-2(H)-chinolinon, TRC
Specifications
Specifications
| Chemischer Name oder Material | 3-Amino-6-nitro-4-phenylquinolin-2(1H)-one |
| CAS | 36020-93-6 |
| Summenformel | C15 H11 N3 O3 |
| Menge | 50 mg |
| Synonym | 3-Amino-6-nitro-4-phenylquinolin-2(1H)-one, Nitrazepam Imp. A (EP) |
| InChI-Formel | InChI=1S/C15H11N3O3/c16-14-13(9-4-2-1-3-5-9)11-8-10(18(20)21)6-7-12(11)17-15(14)19/h1-8H,16H2,(H,17,19) |
| SMILES | NC1=C(c2ccccc2)c3cc(ccc3NC1=O)[N+](=O)[O-] |
| IUPAC-Name | 3-Amino-6-nitro-4-phenyl-1H-chinolin-2-on |
| Molekulargewicht (g/mol) | 281.27 |
| Formelmasse | 281.08 |