missing translation for 'onlineSavingsMsg'
Learn More

3-Aminocarbonyl-5-nitrobenzoesäure, 97 %, Thermo Scientific™

Catalog Number 11336669
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
90196-48-8
C8H6N2O5
210.145
MFCD00127687
BDXFKMQTIGVLEU-UHFFFAOYSA-N
3-aminocarbonyl-5-nitrobenzoic acid, 5-nitroisophthalamic acid, 3-carboxy-5-nitrobenzamide
22749366
3-Carbamoyl-5-Nitrobenzoesäure
C1=C(C=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
90196-48-8
C8H6N2O5
210.145
MFCD00127687
BDXFKMQTIGVLEU-UHFFFAOYSA-N
3-aminocarbonyl-5-nitrobenzoic acid, 5-nitroisophthalamic acid, 3-carboxy-5-nitrobenzamide
22749366
3-Carbamoyl-5-Nitrobenzoesäure
C1=C(C=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Aminocarbonyl-5-nitrobenzoic acid
Schmelzpunkt 239°C to 242°C
Menge 1 g
Formelmasse 210.15
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'