missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylbenzenboronsäure-Pinacolester, 97 %, Thermo Scientific Chemicals

Catalog Number p-7030429
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1073355-05-1
C19H23BO2
294.201
MFCD09266179
VNZSTZMLJUPNQM-UHFFFAOYSA-N
2-3-benzylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-benzylphenylboronic acid pinacol ester, 3-benzylbenzeneboronic acid pinacol ester, amtb038
46739037
2-(3-benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolan
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CC3=CC=CC=C3
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1073355-05-1
C19H23BO2
294.201
MFCD09266179
VNZSTZMLJUPNQM-UHFFFAOYSA-N
2-3-benzylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-benzylphenylboronic acid pinacol ester, 3-benzylbenzeneboronic acid pinacol ester, amtb038
46739037
2-(3-benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolan
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CC3=CC=CC=C3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Benzylbenzeneboronic acid pinacol ester
Schmelzpunkt 68°C to 69°C
Menge 1 g
Formelmasse 294.2
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'