missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzylalkohol, 97%, Thermo Scientific Chemicals

Catalog Number p-7030160
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1700-30-7
C14H14O2
214.264
MFCD00004642
AFKLSWIRJUJWKY-UHFFFAOYSA-N
3-benzyloxybenzyl alcohol, 3-benzyloxy phenyl methanol, 3-benzyloxybenzylalcohol, benzenemethanol, 3-phenylmethoxy, 3-phenylmethoxyphenyl methanol, 3-benzyloxyphenyl methanol, 3-phenylmethoxy phenyl methan-1-ol, pubchem7731, acmc-1bo1a
74341
(3-Phenylmethoxyphenyl)methanol
C1=CC=C(C=C1)COC2=CC=CC(=C2)CO
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1700-30-7
C14H14O2
214.264
MFCD00004642
AFKLSWIRJUJWKY-UHFFFAOYSA-N
3-benzyloxybenzyl alcohol, 3-benzyloxy phenyl methanol, 3-benzyloxybenzylalcohol, benzenemethanol, 3-phenylmethoxy, 3-phenylmethoxyphenyl methanol, 3-benzyloxyphenyl methanol, 3-phenylmethoxy phenyl methan-1-ol, pubchem7731, acmc-1bo1a
74341
(3-Phenylmethoxyphenyl)methanol
C1=CC=C(C=C1)COC2=CC=CC(=C2)CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Benzyloxybenzyl alcohol
Schmelzpunkt 47°C to 51°C
Flammpunkt >110°C (230°F)
Menge 5 g
Formelmasse 214.26
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'