missing translation for 'onlineSavingsMsg'
Learn More

3-Brom-1,1,1-Trifluor-2-Propanol, 96 %, Thermo Scientific Chemicals

Catalog Number p-7045399
Click to view available options
Menge:
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
431-34-5
C3H4BrF3O
192.96
MFCD00040945
VBHIIZIQRDVGDH-UHFFFAOYNA-N
220632
3-Brom-1,1,1-Trifluorpropan-2-ol
OC(CBr)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
431-34-5
C3H4BrF3O
192.96
MFCD00040945
VBHIIZIQRDVGDH-UHFFFAOYNA-N
220632
3-Brom-1,1,1-Trifluorpropan-2-ol
OC(CBr)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Bromo-1,1,1-trifluoro-2-propanol
Dichte 1.861
Siedepunkt 124°C to 126°C
Flammpunkt 43°C (109°F)
Brechungsindex 1.399
Menge 1 g
UN-Nummer UN1987
Beilstein 1737803
Synonym 3-bromo-1,1,1-trifluoro-2-propanol, pubchem12626, acmc-20an9c, 1,1,1-trifluoro-3-bromo-propan-2-ol, 2-propanol,3-bromo-1,1,1-trifluoro, 3-bromo-1,1,1-trifluoro-propan-2-ol, 2-propanol, 3-bromo-1,1,1-trifluoro, rac-3-bromo-1,1,1-trifluoro-propan-2-ol, 3-bromanyl-1,1,1-tris fluoranyl propan-2-ol
Formelmasse 192.97
Reinheit (%) 96%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'