missing translation for 'onlineSavingsMsg'
Learn More

3-Brom-2-nitropyridin, ≥ 98 %, Thermo Scientific Chemicals

Catalog Number p-7047619
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
54231-33-3
C5H3BrN2O2
203.00
MFCD00955614
WFNISJZUJCKTLT-UHFFFAOYSA-N
2-nitro-3-bromopyridine, pyridine, 3-bromo-2-nitro, 3-bromo-2-nitro-pyridine, pubchem4502, pubchem10100, acmc-1ay9l, #, ksc494a7n, 3-bromo-2-nitriopyridine
594044
3-Brom-2-Nitropyridin
[O-][N+](=O)C1=NC=CC=C1Br
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
54231-33-3
C5H3BrN2O2
203.00
MFCD00955614
WFNISJZUJCKTLT-UHFFFAOYSA-N
2-nitro-3-bromopyridine, pyridine, 3-bromo-2-nitro, 3-bromo-2-nitro-pyridine, pubchem4502, pubchem10100, acmc-1ay9l, #, ksc494a7n, 3-bromo-2-nitriopyridine
594044
3-Brom-2-Nitropyridin
[O-][N+](=O)C1=NC=CC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Bromo-2-nitropyridine
Schmelzpunkt 102°C to 106°C
Menge 1 g
Beilstein 1637071
Formelmasse 203
Reinheit (%) ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'