missing translation for 'onlineSavingsMsg'
Learn More

3-Brom-5-Methylbenzotrifluorid, 99 %, Thermo Scientific™

Catalog Number p-7031092
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
86845-28-5
C8H6BrF3
239.04
MFCD01631585
ZDAIYLXHTGIUGY-UHFFFAOYSA-N
3-bromo-5-trifluoromethyl toluene, 1-bromo-3-methyl-5-trifluoromethyl benzene, 3-bromo-5-methylbenzotrifluoride, 1-bromo-3-methyl-5-trifluoromethyl-benzene, 3-methyl-5-trifluoromethyl bromobenzene, 3-bromo-5-methyl-1-trifluoromethyl benzene, benzene,1-bromo-3-methyl-5-trifluoromethyl, 5-bromo-alpha,alpha,alpha-trifluoro-m-xylene, 3-bromo-5-trifluoromethyl toluene 3-bromo-5-methylbenzotrifluoride
20467615
1-bromo-3-methyl-5-(trifluoromethyl)benzene
CC1=CC(=CC(Br)=C1)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
86845-28-5
C8H6BrF3
239.04
MFCD01631585
ZDAIYLXHTGIUGY-UHFFFAOYSA-N
3-bromo-5-trifluoromethyl toluene, 1-bromo-3-methyl-5-trifluoromethyl benzene, 3-bromo-5-methylbenzotrifluoride, 1-bromo-3-methyl-5-trifluoromethyl-benzene, 3-methyl-5-trifluoromethyl bromobenzene, 3-bromo-5-methyl-1-trifluoromethyl benzene, benzene,1-bromo-3-methyl-5-trifluoromethyl, 5-bromo-alpha,alpha,alpha-trifluoro-m-xylene, 3-bromo-5-trifluoromethyl toluene 3-bromo-5-methylbenzotrifluoride
20467615
1-bromo-3-methyl-5-(trifluoromethyl)benzene
CC1=CC(=CC(Br)=C1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Bromo-5-methylbenzotrifluoride
Menge 1 g
Formelmasse 239.04
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'