missing translation for 'onlineSavingsMsg'
Learn More

3-Brombenzenboronsäure, 98+ %, Thermo Scientific Chemicals

Catalog Number p-7046797
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
89598-96-9
C6H6BBrO2
200.83
MFCD00239386
AFSSVCNPDKKSRR-UHFFFAOYSA-N
3-bromophenyl boronic acid, 3-bromobenzeneboronic acid, 3-bromophenylboronicacid, boronic acid, 3-bromophenyl, pubchem1758, 3-bromo-phenylboronic acid, acmc-209r1w, 3-bromo-benzeneboronic acid, 3-bromobenzene boronic acid
2734318
(3-bromophenyl)boronic acid
OB(O)C1=CC=CC(Br)=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
89598-96-9
C6H6BBrO2
200.83
MFCD00239386
AFSSVCNPDKKSRR-UHFFFAOYSA-N
3-bromophenyl boronic acid, 3-bromobenzeneboronic acid, 3-bromophenylboronicacid, boronic acid, 3-bromophenyl, pubchem1758, 3-bromo-phenylboronic acid, acmc-209r1w, 3-bromo-benzeneboronic acid, 3-bromobenzene boronic acid
2734318
(3-bromophenyl)boronic acid
OB(O)C1=CC=CC(Br)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Bromobenzeneboronic acid
Schmelzpunkt 164°C to 168°C
Menge 1 g
Beilstein 3127104
Formelmasse 200.83
Reinheit (%) ≥98%

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'