missing translation for 'onlineSavingsMsg'
Learn More

3-Chlor-2,6-Difluorbenzoylchlorid, 97 %, Thermo Scientific™

Catalog Number p-7047240
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
261762-43-0
C7H2Cl2F2O
210.989
MFCD01631324
LUTDLSSUGZTZBP-UHFFFAOYSA-N
3-chloro-2,6-difluorobenzoylchloride, acmc-1cff7, 3-chloro-2,6-difluoro-benzoyl chloride, benzoyl chloride,3-chloro-2,6-difluoro, 3-chloranyl-2,6-bis fluoranyl benzoyl chloride, benzoyl chloride, 3-chloro-2,6-difluoro-9ci
2773540
3-Chlor-2,6-difluorbenzoylchlorid
C1=CC(=C(C(=C1F)C(=O)Cl)F)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
261762-43-0
C7H2Cl2F2O
210.989
MFCD01631324
LUTDLSSUGZTZBP-UHFFFAOYSA-N
3-chloro-2,6-difluorobenzoylchloride, acmc-1cff7, 3-chloro-2,6-difluoro-benzoyl chloride, benzoyl chloride,3-chloro-2,6-difluoro, 3-chloranyl-2,6-bis fluoranyl benzoyl chloride, benzoyl chloride, 3-chloro-2,6-difluoro-9ci
2773540
3-Chlor-2,6-difluorbenzoylchlorid
C1=CC(=C(C(=C1F)C(=O)Cl)F)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Chloro-2,6-difluorobenzoyl chloride
Brechungsindex 1.52
Menge 1 g
UN-Nummer UN3265
Empfindlichkeit Moisture sensitive
Formelmasse 211
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'