missing translation for 'onlineSavingsMsg'
Learn More

3-Chlor-2-methoxypyridin-4-Boronsäure-Pinacolester, 95 %, Thermo Scientific™

Catalog Number p-7029988
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1073353-73-7
C12H17BClNO3
269.53
MFCD06798258
JYVZFQGNBUYGIH-UHFFFAOYSA-N
3-chloro-2-methoxypyridine-4-boronic acid pinacol ester, 3-chloro-2-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-chloro-2-methoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 3-chloro-2-methoxy-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-chloro-2-methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-chloro-2-methoxypyridine-4-boronic acid, pinacol ester, pyridine, 3-chloro-2-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
44755159
3-Chlor-2-Methoxy-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)pyridin
COC1=NC=CC(B2OC(C)(C)C(C)(C)O2)=C1Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1073353-73-7
C12H17BClNO3
269.53
MFCD06798258
JYVZFQGNBUYGIH-UHFFFAOYSA-N
3-chloro-2-methoxypyridine-4-boronic acid pinacol ester, 3-chloro-2-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-chloro-2-methoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 3-chloro-2-methoxy-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-chloro-2-methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 3-chloro-2-methoxypyridine-4-boronic acid, pinacol ester, pyridine, 3-chloro-2-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
44755159
3-Chlor-2-Methoxy-4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)pyridin
COC1=NC=CC(B2OC(C)(C)C(C)(C)O2)=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Chloro-2-methoxypyridine-4-boronic acid pinacol ester
Schmelzpunkt 47°C to 50°C
Menge 250 mg
Formelmasse 269.53
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'