missing translation for 'onlineSavingsMsg'
Learn More

3-Chlor-4-(3-pyridyl)-1,2,5-thiadiazol, 95 %, Thermo Scientific Chemicals

Catalog Number p-7031188
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
131986-28-2
C7H4ClN3S
197.64
MFCD03066187
CMPNWGQBNRHIQZ-UHFFFAOYSA-N
3-chloro-4-pyridin-3-yl-1,2,5-thiadiazole, 3-chloro-4-3-pyridyl-1,2,5-thiadiazole, 3-4-chloro-1,2,5-thiadiazol-3-yl pyridine, pyridine, 3-4-chloro-1,2,5-thiadiazol-3-yl, 3-4-chloro-1,2,5 thiadiazol-3-yl-pyridine, 3-3-chloro-1,2,5-thiadiazol-4yl pyridine, 3-3-chloro-1,2,5-thiadiazol-4-yl pyridine, acmc-1bz6y, ksc497a5t, 3-chloro-4-pyrid-3-yl-1,2,5-thiadiazole
10488164
3-Chlor-4-Pyridin-3-yl-1,2,5-Thiadiazol
ClC1=NSN=C1C1=CN=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
131986-28-2
C7H4ClN3S
197.64
MFCD03066187
CMPNWGQBNRHIQZ-UHFFFAOYSA-N
3-chloro-4-pyridin-3-yl-1,2,5-thiadiazole, 3-chloro-4-3-pyridyl-1,2,5-thiadiazole, 3-4-chloro-1,2,5-thiadiazol-3-yl pyridine, pyridine, 3-4-chloro-1,2,5-thiadiazol-3-yl, 3-4-chloro-1,2,5 thiadiazol-3-yl-pyridine, 3-3-chloro-1,2,5-thiadiazol-4yl pyridine, 3-3-chloro-1,2,5-thiadiazol-4-yl pyridine, acmc-1bz6y, ksc497a5t, 3-chloro-4-pyrid-3-yl-1,2,5-thiadiazole
10488164
3-Chlor-4-Pyridin-3-yl-1,2,5-Thiadiazol
ClC1=NSN=C1C1=CN=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Chloro-4-(3-pyridyl)-1,2,5-thiadiazole
Schmelzpunkt 56°C
Menge 250 mg
Formelmasse 197.65
Reinheit (%) 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'