missing translation for 'onlineSavingsMsg'
Learn More

3'-Chlor-4'-Ethoxy-5'-Fluoroacetophenon, 97 %, Thermo Scientific™

Catalog Number p-7031461
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1017779-05-3
C10H10ClFO2
216.636
MFCD09832347
NXUGPYUMPWKWQU-UHFFFAOYSA-N
3'-chloro-4'-ethoxy-5'-fluoroacetophenone, 1-3-chloro-4-ethoxy-5-fluorophenyl ethanone, 1-acetyl-3-chloro-4-ethoxy-5-fluorobenzene, 3'-chloro-4'-ethoxy-5'-fluoroacetophenone, jrd
46737663
1-(3-Chlor-4-Ethoxy-5-Fluorphenyl)ethanon
CCOC1=C(C=C(C=C1Cl)C(=O)C)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1017779-05-3
C10H10ClFO2
216.636
MFCD09832347
NXUGPYUMPWKWQU-UHFFFAOYSA-N
3'-chloro-4'-ethoxy-5'-fluoroacetophenone, 1-3-chloro-4-ethoxy-5-fluorophenyl ethanone, 1-acetyl-3-chloro-4-ethoxy-5-fluorobenzene, 3'-chloro-4'-ethoxy-5'-fluoroacetophenone, jrd
46737663
1-(3-Chlor-4-Ethoxy-5-Fluorphenyl)ethanon
CCOC1=C(C=C(C=C1Cl)C(=O)C)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3'-Chloro-4'-ethoxy-5'-fluoroacetophenone
Brechungsindex 1.526
Menge 1 g
Formelmasse 216.64
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'