missing translation for 'onlineSavingsMsg'
Learn More

3-Chlor-5-(Pentafluorothio)Benzamid, 97 %

Catalog Number p-7031894
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1240257-35-5
C7H5ClF5NOS
281.63
MFCD16652452
SCYISRHUJQHNFD-UHFFFAOYSA-N
3-chloro-5-pentafluorosulfur benzamide, 3-chloro-5-pentafluoro-??-sulfanyl benzamide, 3-chloro-5-pentafluorothio benzamide
66523520
3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzamide
NC(=O)C1=CC(=CC(Cl)=C1)S(F)(F)(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1240257-35-5
C7H5ClF5NOS
281.63
MFCD16652452
SCYISRHUJQHNFD-UHFFFAOYSA-N
3-chloro-5-pentafluorosulfur benzamide, 3-chloro-5-pentafluoro-??-sulfanyl benzamide, 3-chloro-5-pentafluorothio benzamide
66523520
3-chloro-5-(pentafluoro-λ⁶-sulfanyl)benzamide
NC(=O)C1=CC(=CC(Cl)=C1)S(F)(F)(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Chloro-5-(pentafluorothio)benzamide
Menge 250 mg
Formelmasse 281.63
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'