missing translation for 'onlineSavingsMsg'
Learn More

3-Chlor-5-(pentafluorthio)benzylalkohol, 97 %

Catalog Number 15466297
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1240256-97-6
C7H6ClF5OS
268.63
MFCD16652422
JXKDNNYDGFDTFO-UHFFFAOYSA-N
3-chloro-5-pentafluorosulfur benzyl alcohol, 3-chloro-5-pentafluoro-??-sulfanyl phenyl methanol, 3-chloro-5-pentafluorothio benzyl alcohol
66523462
[3-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]methanol
OCC1=CC(=CC(Cl)=C1)S(F)(F)(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1240256-97-6
C7H6ClF5OS
268.63
MFCD16652422
JXKDNNYDGFDTFO-UHFFFAOYSA-N
3-chloro-5-pentafluorosulfur benzyl alcohol, 3-chloro-5-pentafluoro-??-sulfanyl phenyl methanol, 3-chloro-5-pentafluorothio benzyl alcohol
66523462
[3-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]methanol
OCC1=CC(=CC(Cl)=C1)S(F)(F)(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Chloro-5-(pentafluorothio)benzyl alcohol
Menge 1 g
Formelmasse 268.63
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'