missing translation for 'onlineSavingsMsg'
Learn More

3-Chlor-5-(Trifluormethyl)pyridin-2-Carbonsäure, 97 %, Thermo Scientific Chemicals

Catalog Number p-7032695
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
80194-68-9
C7H3ClF3NO2
225.551
MFCD00277482
HXRMCZBDTDCCOP-UHFFFAOYSA-N
3-chloro-5-trifluoromethyl pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl picolinic acid, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl-2-pyridinecarboxylic acid, 2-carboxy-3-chloro-5-trifluoromethyl pyridine, ksc446c7n, 3-chloro-5-trifluoromethylpicolinic acid, 3-chloro-5-trifluoromethyl-picolinic acid
2821908
3-Chlor-5-(Trifluormethyl)pyridin-2-Carbonsäure
C1=C(C=NC(=C1Cl)C(=O)O)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
80194-68-9
C7H3ClF3NO2
225.551
MFCD00277482
HXRMCZBDTDCCOP-UHFFFAOYSA-N
3-chloro-5-trifluoromethyl pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl picolinic acid, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl, 3-chloro-5-trifluoromethyl-pyridine-2-carboxylic acid, 3-chloro-5-trifluoromethyl-2-pyridinecarboxylic acid, 2-carboxy-3-chloro-5-trifluoromethyl pyridine, ksc446c7n, 3-chloro-5-trifluoromethylpicolinic acid, 3-chloro-5-trifluoromethyl-picolinic acid
2821908
3-Chlor-5-(Trifluormethyl)pyridin-2-Carbonsäure
C1=C(C=NC(=C1Cl)C(=O)O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid
Schmelzpunkt 141°C to 143°C
Menge 250 mg
Formelmasse 225.6
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'