Learn More
3-Chlorobenzotrifluoride, TRC
Product Code. 30362911
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362911 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30362911 Supplier Toronto Research Chemicals Supplier No. TRCC42874550G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Chloro-alpha,alpha,alpha-trifluorotoluene
CAS 98-15-7
Molecular Formula C7 H4 Cl F3
Quantity 50 g
Synonym Fluoxetine Impurity 16
InChI Formula InChI=1S/C7H4ClF3/c8-6-3-1-2-5(4-6)7(9,10)11/h1-4H
SMILES FC(F)(F)c1cccc(Cl)c1
IUPAC Name 1-chloro-3-(trifluoromethyl)benzene
Molecular Weight (g/mol) 180.55
Formula Weight 179.9954