missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-Methylpyridin, 97 %, Thermo Scientific Chemicals

Catalog Number p-7047798
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5444-01-9
MFCD00234272
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5444-01-9
MFCD00234272

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Cyano-4-methylpyridine
Schmelzpunkt 41°C to 44°C
Siedepunkt 108°C to 111°C (20 mmHg)
Geruch Characteristic
Summenformel C7H6N2
Menge 1 g
Beilstein 112016
Synonym 3-cyano-4-methylpyridine, 4-methylnicotinonitrile, 3-cyano-4-picoline, 4-methyl-nicotinonitrile, 3-pyridinecarbonitrile, 4-methyl, 4-methyl-3-pyridinecarbonitrile, 3-cyano-4-methylpyridine 3-cyano-4-picoline, pubchem6623, pubchem18426, 4-methyl nicotinonitrile
InChI-Schlüssel XLAPHZHNODDMDD-UHFFFAOYSA-N
SMILES CC1=C(C=NC=C1)C#N
IUPAC-Name 4-Methylpyridin-3-Carbonitril
Molekulargewicht (g/mol) 118.14
PubChem CID 227806
Formelmasse 118.14
Reinheit (%) 97%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'