missing translation for 'onlineSavingsMsg'
Learn More

3-Di-n-propylcarbamoyl-4-Fluorobenzolboronsäure, 97 %, Thermo Scientific™

Catalog Number p-7036660
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2096330-04-8
C13H19BFNO3
267.11
MFCD20134177
ZOWSBHXCHLRWLC-UHFFFAOYSA-N
3-di-n-propylcarbamoyl-4-fluorobenzeneboronic acid
73996066
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2096330-04-8
C13H19BFNO3
267.11
MFCD20134177
ZOWSBHXCHLRWLC-UHFFFAOYSA-N
3-di-n-propylcarbamoyl-4-fluorobenzeneboronic acid
73996066

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Di-n-propylcarbamoyl-4-fluorobenzeneboronic acid
Menge 250 mg
Formelmasse 267.1
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'