Learn More
3-Dimethylamino-1-(2-methoxyphenyl)-2-propen-1-on, 95 %, Thermo Scientific™
Catalog Number 15468315
Click to view available options
Menge:
5 g
25 g
195813-59-3
C12H15NO2
205.257
MFCD08689452
WKNJWYUQADHZSM-CMDGGOBGSA-N
2e-3-dimethylamino-1-2-methoxyphenyl prop-2-en-1-one, 2e-3-dimethylamino-1-2-methoxyphenyl-2-propen-1-one, 2e-3-dimethylamino-1-2-methoxyphenyl-prop-2-en-1-one, 3-dimethylamino-1-2-methoxyphenyl-2-propen-1-one, 2-propen-1-one, 3-dimethylamino-1-2-methoxyphenyl-, 2e
10465548
(E)-3-(dimethylamino)-1-(2-methoxyphenyl)prop-2-en-1-on
CN(C)C=CC(=O)C1=CC=CC=C1OC
195813-59-3
C12H15NO2
205.257
MFCD08689452
WKNJWYUQADHZSM-CMDGGOBGSA-N
2e-3-dimethylamino-1-2-methoxyphenyl prop-2-en-1-one, 2e-3-dimethylamino-1-2-methoxyphenyl-2-propen-1-one, 2e-3-dimethylamino-1-2-methoxyphenyl-prop-2-en-1-one, 3-dimethylamino-1-2-methoxyphenyl-2-propen-1-one, 2-propen-1-one, 3-dimethylamino-1-2-methoxyphenyl-, 2e
10465548
(E)-3-(dimethylamino)-1-(2-methoxyphenyl)prop-2-en-1-on
CN(C)C=CC(=O)C1=CC=CC=C1OC
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Dimethylamino-1-(2-methoxyphenyl)-2-propen-1-one
Schmelzpunkt 59°C to 63°C
Menge 5 g
Formelmasse 205.26
Reinheit (%) 95%