missing translation for 'onlineSavingsMsg'
Learn More

3-Fluor-2-Formylbenzolboronsäure-Pinacolester, 96 %, Thermo Scientific™

Catalog Number 15407896
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1246633-35-1
C13H16BFO3
250.08
PMDUBXLKNQBIHI-UHFFFAOYSA-N
2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-6-fluorobenzaldehyde
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1246633-35-1
C13H16BFO3
250.08
PMDUBXLKNQBIHI-UHFFFAOYSA-N
2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-6-fluorobenzaldehyde

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Fluoro-2-formylbenzeneboronic acid pinacol ester
Menge 1 g
Formelmasse 250.07
Reinheit (%) 96%
Physikalische Form Fest
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'