missing translation for 'onlineSavingsMsg'
Learn More

3-Fluor-5-(pentafluorthio)-Benzoesäure, 97 %, Thermo Scientific™

Catalog Number p-7032702
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1448319-13-8
C7H4F6O2S
266.157
MFCD16652421
WQRFQFPUMUJGOL-UHFFFAOYSA-N
3-fluoro-5-pentafluorosulfur benzoic acid, 3-fluoro-5-pentafluoro-??-sulfanyl benzoic acid, 3-fluoro-5-pentafluorothio benzoic acid, 3-fluoro-5-pentafluoro-lambda6-sulphanyl benzoic acid
66523461
3-fluor-5-(pentafluor-$l^{6}-sulfanyl)benzoesäure
C1=C(C=C(C=C1F)S(F)(F)(F)(F)F)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1448319-13-8
C7H4F6O2S
266.157
MFCD16652421
WQRFQFPUMUJGOL-UHFFFAOYSA-N
3-fluoro-5-pentafluorosulfur benzoic acid, 3-fluoro-5-pentafluoro-??-sulfanyl benzoic acid, 3-fluoro-5-pentafluorothio benzoic acid, 3-fluoro-5-pentafluoro-lambda6-sulphanyl benzoic acid
66523461
3-fluor-5-(pentafluor-$l^{6}-sulfanyl)benzoesäure
C1=C(C=C(C=C1F)S(F)(F)(F)(F)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Fluoro-5-(pentafluorothio)benzoic acid
Menge 250 mg
Formelmasse 266.16
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'