missing translation for 'onlineSavingsMsg'
Learn More

3-Fluor-5-(trifluormethoxy)benzylamin, 97 %, Thermo Scientific™

Catalog Number p-7032642
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1353000-19-7
C8H7F4NO
209.144
MFCD18783230
ULLWGZMNUJLDKA-UHFFFAOYSA-N
3-fluoro-5-trifluoromethoxy benzylamine, 3-fluoro-5-trifluoromethoxy phenyl methanamine, 1-3-fluoro-5-trifluoromethoxy phenyl methanamine
66523552
[3-fluor-5-(trifluormethoxy)phenyl]methanamin
C1=C(C=C(C=C1OC(F)(F)F)F)CN
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1353000-19-7
C8H7F4NO
209.144
MFCD18783230
ULLWGZMNUJLDKA-UHFFFAOYSA-N
3-fluoro-5-trifluoromethoxy benzylamine, 3-fluoro-5-trifluoromethoxy phenyl methanamine, 1-3-fluoro-5-trifluoromethoxy phenyl methanamine
66523552
[3-fluor-5-(trifluormethoxy)phenyl]methanamin
C1=C(C=C(C=C1OC(F)(F)F)F)CN

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Fluoro-5-(trifluoromethoxy)benzylamine
Brechungsindex 1.4395
Menge 250 mg
UN-Nummer UN3259
Empfindlichkeit Air Sensitive
Formelmasse 209.14
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'