missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorosalicylsäure, 99 %, Thermo Scientific™

Catalog Number p-7028756
Click to view available options
Menge:
500 mg
2 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
341-27-5
C7H5FO3
156.112
MFCD00153167
GFHCXVJXSLGRJR-UHFFFAOYSA-N
3-fluorosalicylic acid, benzoic acid, 3-fluoro-2-hydroxy, 2-hydroxy-3-fluorobenzoic acid, 3-fluoro-2-hydroxy-benzoic acid, salicylic acid, 3-fluoro, 2-hydroxy-3-fluorobenzoicacid, 3-fluoro-2-hydroxybenzoicacid, pubchem4960, acmc-1cjoo
268724
3-Fluor-2-hydroxybenzoesäure
C1=CC(=C(C(=C1)F)O)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
341-27-5
C7H5FO3
156.112
MFCD00153167
GFHCXVJXSLGRJR-UHFFFAOYSA-N
3-fluorosalicylic acid, benzoic acid, 3-fluoro-2-hydroxy, 2-hydroxy-3-fluorobenzoic acid, 3-fluoro-2-hydroxy-benzoic acid, salicylic acid, 3-fluoro, 2-hydroxy-3-fluorobenzoicacid, 3-fluoro-2-hydroxybenzoicacid, pubchem4960, acmc-1cjoo
268724
3-Fluor-2-hydroxybenzoesäure
C1=CC(=C(C(=C1)F)O)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Fluorosalicylic acid
Schmelzpunkt 146°C to 151°C
Menge 500 mg
Formelmasse 156.11
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'