missing translation for 'onlineSavingsMsg'
Learn More

3-Hexanol, 98%

Product Code. 11409666
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11409666

Brand: Thermo Scientific Alfa Aesar B20210.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-37-0 | C6H14O | 102.177 g/mol

It finds it application as a flavoring agent or adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it application as a flavoring agent or adjuvant.

Solubility
Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-37-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004582
InChI Key ZOCHHNOQQHDWHG-UHFFFAOYSA-N
Synonym 3-hexanol, ethyl propyl carbinol, 3-hexyl alcohol, 3-hexanol natural, ethylpropylcarbinol, fema no. 3351, hexanol-3, 1-butanol, ethyl, acmc-209n2d
PubChem CID 12178
IUPAC Name hexan-3-ol
SMILES CCCC(CC)O

Specifications

Density 0.81
Boiling Point 134°C to 136°C
Flash Point 42°C (108°F)
Odor Characteristic
Refractive Index 1.415
Quantity 100 g
UN Number UN2282
Beilstein 1718964
Solubility Information Soluble in alcohol.
Formula Weight 102.18
Percent Purity 98%
Chemical Name or Material 3-Hexanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.