missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-3-methyl-2-butanonoxim, 98 %, Thermo Scientific™

Catalog Number p-7045416
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7431-25-6
C5H11NO2
117.148
MFCD00059656
QKLLBCGVADVPKD-XQRVVYSFSA-N
3-hydroxy-3-methyl-2-butanone oxime, 3e-3-hydroxyimino-2-methylbutan-2-ol, qkllbcgvadvpkd-xqrvvysfsa, 3-hydroxy-3-methyl-butan-2-one oxime, 3z-3-hydroxyimino-2-methylbutan-2-ol, 3e-3-hydroxyimino-2-methyl-butan-2-ol, 2z-3-hydroxy-3-methyl-2-butanone oxime #
5385758
(3Z)-3-Hydroxyimino-2-Methylbutan-2-ol
CC(=NO)C(C)(C)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7431-25-6
C5H11NO2
117.148
MFCD00059656
QKLLBCGVADVPKD-XQRVVYSFSA-N
3-hydroxy-3-methyl-2-butanone oxime, 3e-3-hydroxyimino-2-methylbutan-2-ol, qkllbcgvadvpkd-xqrvvysfsa, 3-hydroxy-3-methyl-butan-2-one oxime, 3z-3-hydroxyimino-2-methylbutan-2-ol, 3e-3-hydroxyimino-2-methyl-butan-2-ol, 2z-3-hydroxy-3-methyl-2-butanone oxime #
5385758
(3Z)-3-Hydroxyimino-2-Methylbutan-2-ol
CC(=NO)C(C)(C)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Hydroxy-3-methyl-2-butanone oxime
Schmelzpunkt 84°C to 86°C
Menge 5 g
Beilstein 1747660
Formelmasse 117.15
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'