missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-(2,2,2-trimethylacetamido)pyridin, 98 %, Thermo Scientific™

Catalog Number p-7033030
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
169205-93-0
C10H14N2O2
194.234
MFCD07776841
HZNFSZMIMUTCGG-UHFFFAOYSA-N
n-3-hydroxypyridin-4-yl-2,2-dimethylpropanamide, n-3-hydroxy-pyridin-4-yl-2,2-dimethyl-propionamide, n-3-hydroxypyridin-4-yl pivalamide, 3-hydroxy-4-2,2,2-trimethylacetamido pyridine, n-3-hydroxy 4-pyridyl-2,2-dimethylpropanamide, acmc-1bzqc, 2,2-dimethyl-n-3-oxidanylpyridin-4-yl propanamide, n-3-hydroxy-4-pyridinyl-2,2-dimethylpropanamide, n-3-hydroxy-4-pyridinyl-2,2-dimethyl-propionamide, propanamide,n-3-hydroxy-4-pyridinyl-2,2-dimethyl
10035519
N-(3-Hydroxypyridin-4-yl)-2,2-dimethylpropanamid
CC(C)(C)C(=O)NC1=C(C=NC=C1)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
169205-93-0
C10H14N2O2
194.234
MFCD07776841
HZNFSZMIMUTCGG-UHFFFAOYSA-N
n-3-hydroxypyridin-4-yl-2,2-dimethylpropanamide, n-3-hydroxy-pyridin-4-yl-2,2-dimethyl-propionamide, n-3-hydroxypyridin-4-yl pivalamide, 3-hydroxy-4-2,2,2-trimethylacetamido pyridine, n-3-hydroxy 4-pyridyl-2,2-dimethylpropanamide, acmc-1bzqc, 2,2-dimethyl-n-3-oxidanylpyridin-4-yl propanamide, n-3-hydroxy-4-pyridinyl-2,2-dimethylpropanamide, n-3-hydroxy-4-pyridinyl-2,2-dimethyl-propionamide, propanamide,n-3-hydroxy-4-pyridinyl-2,2-dimethyl
10035519
N-(3-Hydroxypyridin-4-yl)-2,2-dimethylpropanamid
CC(C)(C)C(=O)NC1=C(C=NC=C1)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Hydroxy-4-(2,2,2-trimethylacetamido)pyridine
Menge 250 mg
Formelmasse 194.24
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'