missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4'-Methoxyflavon, 97 %, Thermo Scientific™

Catalog Number p-7029842
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
6889-78-7
C16H12O4
268.268
MFCD00017682
IIBBFGMVMNZMGA-UHFFFAOYSA-N
4'-methoxyflavonol, 3-hydroxy-4'-methoxyflavone, 3-hydroxy-2-4-methoxyphenyl-4h-chromen-4-one, 3-hydroxy-2-4-methoxyphenyl chromen-4-one, 4h-1-benzopyran-4-one, 3-hydroxy-2-4-methoxyphenyl, 2-4-methoxyphenyl-3-hydroxyflavone, hydroxymethoxyphenylchromenone, 3-hydroxy-4'-methoxyflavanone, 4h-1-benzopyran-4-one, 3-hydroxy-2-4-methoxyphenyl-9ci
97141
3-Hydroxy-2-(4-methoxyphenyl)chromen-4-on
COC1=CC=C(C=C1)C2=C(C(=O)C3=CC=CC=C3O2)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
6889-78-7
C16H12O4
268.268
MFCD00017682
IIBBFGMVMNZMGA-UHFFFAOYSA-N
4'-methoxyflavonol, 3-hydroxy-4'-methoxyflavone, 3-hydroxy-2-4-methoxyphenyl-4h-chromen-4-one, 3-hydroxy-2-4-methoxyphenyl chromen-4-one, 4h-1-benzopyran-4-one, 3-hydroxy-2-4-methoxyphenyl, 2-4-methoxyphenyl-3-hydroxyflavone, hydroxymethoxyphenylchromenone, 3-hydroxy-4'-methoxyflavanone, 4h-1-benzopyran-4-one, 3-hydroxy-2-4-methoxyphenyl-9ci
97141
3-Hydroxy-2-(4-methoxyphenyl)chromen-4-on
COC1=CC=C(C=C1)C2=C(C(=O)C3=CC=CC=C3O2)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Hydroxy-4'-methoxyflavone
Menge 1 g
Formelmasse 268.26
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'