missing translation for 'onlineSavingsMsg'
Learn More

3-(Hydroxymethyl)-Benzolboronsäure-Pinacolester, Thermo Scientific Chemicals

Catalog Number p-7033925
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
443776-76-9
C13H19BO3
234.102
MFCD09266196
ZEWWJJQAFTXUIS-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethyl phenylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethylphenylboronic acid pinacol ester, benzenemethanol, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, amtb056, 3-hydroxymethyl benzenboronic acid pinacol ester, 3-hydroxymethylphenylboronic acid, pinacol ester, 3-hydroxymethyl benzeneboronic acid pinacol ester
44118236
[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CO
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
443776-76-9
C13H19BO3
234.102
MFCD09266196
ZEWWJJQAFTXUIS-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethyl phenylboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanol, 3-hydroxymethylphenylboronic acid pinacol ester, benzenemethanol, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, amtb056, 3-hydroxymethyl benzenboronic acid pinacol ester, 3-hydroxymethylphenylboronic acid, pinacol ester, 3-hydroxymethyl benzeneboronic acid pinacol ester
44118236
[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(Hydroxymethyl)benzeneboronic acid pinacol ester
Menge 1 g
Formelmasse 234.1

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'