missing translation for 'onlineSavingsMsg'
Learn More

3-Iod-4-Methoxybenzoesäure, 98 %, Thermo Scientific™

Catalog Number p-7027775
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
68507-19-7
C8H7IO3
278.045
MFCD00017347
HNJSYSWRPCCSQI-UHFFFAOYSA-N
624158
3-Iod-4-methoxybenzoesäure
COC1=C(C=C(C=C1)C(=O)O)I
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
68507-19-7
C8H7IO3
278.045
MFCD00017347
HNJSYSWRPCCSQI-UHFFFAOYSA-N
624158
3-Iod-4-methoxybenzoesäure
COC1=C(C=C(C=C1)C(=O)O)I

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Iodo-4-methoxybenzoic acid
Schmelzpunkt 238°C to 241°C
Menge 1 g
Empfindlichkeit Light sensitive
Synonym 3-iodo-4-methoxy-benzoic acid, 3-iodo-p-anisic acid, benzoic acid, 3-iodo-4-methoxy, 3-iodo-4-methoxybenzoicacid, pubchem4973, acmc-209o3r, akos bbb/204, benzoic acid,3-iodo-4-methoxy, 3-iodo-4-methoxybenzoic acid
Formelmasse 278.05
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'