missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Isocyanato-2-methyl-6-(trifluormethyl)pyridin, Thermo Scientific™
Click to view available options
Menge:
1 g
Spezifikation
Chemischer Name oder Material | 3-Isocyanato-2-methyl-6-(trifluoromethyl)pyridine |
CAS | 874832-10-7 |
Summenformel | C8H5F3N2O |
Menge | 1 g |
Synonym | 3-isocyanato-2-methyl-6-trifluoromethyl pyridine, pyridine, 3-isocyanato-2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethyl pyridin-3-isocyanate |
InChI-Schlüssel | BRXUOLWIERBAPA-UHFFFAOYSA-N |
SMILES | CC1=C(C=CC(=N1)C(F)(F)F)N=C=O |
IUPAC-Name | 3-Isocyanato-2-Methyl-6-(Trifluoromethyl)Pyridin |
Molekulargewicht (g/mol) | 202.136 |
PubChem CID | 45594323 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'