Learn More
3-Methoxy-5-(pentafluorthio)benzyl Alkohol, 97 %, Thermo Scientific™
Catalog Number p-7031468
Click to view available options
Menge:
1 g
5 g
916420-98-9
C9H9F3O2
206.164
MFCD09025413
ROACFQURKLEGKL-UHFFFAOYSA-N
3-methoxy-5-trifluoromethyl phenyl methanol, 3-methoxy-5-trifluoromethyl benzyl alcohol, 3-methoxy-5-trifluoromethyl-phenyl-methanol, benzenemethanol, 3-methoxy-5-trifluoromethyl, 5-methoxy-3-trifluoromethyl phenyl methan-1-ol, 3-trifluoromethyl-5-methoxyphenyl methanol
46737578
[3-methoxy-5-(trifluormethyl)phenyl]methanol
COC1=CC(=CC(=C1)C(F)(F)F)CO
916420-98-9
C9H9F3O2
206.164
MFCD09025413
ROACFQURKLEGKL-UHFFFAOYSA-N
3-methoxy-5-trifluoromethyl phenyl methanol, 3-methoxy-5-trifluoromethyl benzyl alcohol, 3-methoxy-5-trifluoromethyl-phenyl-methanol, benzenemethanol, 3-methoxy-5-trifluoromethyl, 5-methoxy-3-trifluoromethyl phenyl methan-1-ol, 3-trifluoromethyl-5-methoxyphenyl methanol
46737578
[3-methoxy-5-(trifluormethyl)phenyl]methanol
COC1=CC(=CC(=C1)C(F)(F)F)CO
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Methoxy-5-(trifluoromethyl)benzyl alcohol
Schmelzpunkt 32°C to 35°C
Menge 1 g
Formelmasse 206.16
Reinheit (%) 97%