missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzamidoxim, 97 %, Thermo Scientific™

Catalog Number p-7034459
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
73647-50-4
C8H10N2O2
166.18
MFCD07161438,MFCD07161438
CEHMGZTZNNEADY-UHFFFAOYSA-N
z-n'-hydroxy-3-methoxybenzimidamide, 3-methoxybenzamidoxime, n'-hydroxy-3-methoxybenzimidamide, e-n'-hydroxy-3-methoxybenzimidamide, n-hydroxy-3-methoxy-benzamidine, n-hydroxy-3-methoxybenzenecarboximidamide, hydroxyimino 3-methoxyphenyl methylamine, n'-hydroxy-3-methoxybenzene-1-carboximidamide, z-n'-hydroxy-3-methoxybenzene-1-carboximidamide, e-n'-hydroxy-3-methoxybenzenecarboximidamide
9652366
N'-hydroxy-3-methoxybenzene-1-carboximidamide
COC1=CC=CC(=C1)C(N)=NO
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
73647-50-4
C8H10N2O2
166.18
MFCD07161438,MFCD07161438
CEHMGZTZNNEADY-UHFFFAOYSA-N
z-n'-hydroxy-3-methoxybenzimidamide, 3-methoxybenzamidoxime, n'-hydroxy-3-methoxybenzimidamide, e-n'-hydroxy-3-methoxybenzimidamide, n-hydroxy-3-methoxy-benzamidine, n-hydroxy-3-methoxybenzenecarboximidamide, hydroxyimino 3-methoxyphenyl methylamine, n'-hydroxy-3-methoxybenzene-1-carboximidamide, z-n'-hydroxy-3-methoxybenzene-1-carboximidamide, e-n'-hydroxy-3-methoxybenzenecarboximidamide
9652366
N'-hydroxy-3-methoxybenzene-1-carboximidamide
COC1=CC=CC(=C1)C(N)=NO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Methoxybenzamidoxime
Menge 1 g
Formelmasse 166.18
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'