missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-(1-piperazinyl)-chinolin, 97 %, Thermo Scientific™

Catalog Number p-7033683
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
348133-74-4
C14H17N3
227.311
MFCD08061017
YUKANULGPVFVJO-UHFFFAOYSA-N
3-methyl-2-piperazin-1-yl quinoline, 3-methyl-2-1-piperazinyl quinoline, 3-methyl-2-piperazin-1-yl-quinoline, acmc-20an0s, 3-methyl-2-quinolyl piperazine, 3-methyl-2-piperazine-1-yl quinoline
22636341
3-Methyl-2-Piperazin-1-ylchinolin
CC1=CC2=CC=CC=C2N=C1N3CCNCC3
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
348133-74-4
C14H17N3
227.311
MFCD08061017
YUKANULGPVFVJO-UHFFFAOYSA-N
3-methyl-2-piperazin-1-yl quinoline, 3-methyl-2-1-piperazinyl quinoline, 3-methyl-2-piperazin-1-yl-quinoline, acmc-20an0s, 3-methyl-2-quinolyl piperazine, 3-methyl-2-piperazine-1-yl quinoline
22636341
3-Methyl-2-Piperazin-1-ylchinolin
CC1=CC2=CC=CC=C2N=C1N3CCNCC3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Methyl-2-(1-piperazinyl)quinoline
Menge 250 mg
Empfindlichkeit Air Sensitive
Formelmasse 227.31
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'