Learn More
3-Methylphthalsäureanhydrid, 96 %, Thermo Scientific Chemicals
Catalog Number p-7026705
Click to view available options
Menge:
1 g
5 g
25 g
4792-30-7
C9H6O3
162.14
MFCD00047316
TWWAWPHAOPTQEU-UHFFFAOYSA-N
3-methylphthalic anhydride, 4-methylisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-methyl, 3-methyl phthalic anhydride, methylphthalic anhydride, phthalic anhydride, 3-methyl, 2,3-toluenedicarboxylic anhydride, 1,3-isobenzofurandione, methyl, 4-methyl-1,3-dihydro-2-benzofuran-1,3-dione, acmc-1arrb
98500
4-Methyl-2-Benzofuran-1,3-Dion
CC1=CC=CC2=C1C(=O)OC2=O
4792-30-7
C9H6O3
162.14
MFCD00047316
TWWAWPHAOPTQEU-UHFFFAOYSA-N
3-methylphthalic anhydride, 4-methylisobenzofuran-1,3-dione, 1,3-isobenzofurandione, 4-methyl, 3-methyl phthalic anhydride, methylphthalic anhydride, phthalic anhydride, 3-methyl, 2,3-toluenedicarboxylic anhydride, 1,3-isobenzofurandione, methyl, 4-methyl-1,3-dihydro-2-benzofuran-1,3-dione, acmc-1arrb
98500
4-Methyl-2-Benzofuran-1,3-Dion
CC1=CC=CC2=C1C(=O)OC2=O
3-Methylphthalic anhydride can react with Methylamine hydrochloride to get 2,3-Dihydro-2,4-dimethyl-1H-isoindole-1,3-dione.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
3-Methylphthalanhydrid kann mit Methylaminhydrochlorid reagieren, um 2,3-Dihydro-2,4-dimethyl-1H-isoindol-1,3-dion zu erzeugen.
Löslichkeit
Es reagiert mit Wasser.
Hinweise
Feuchtigkeitsempfindlich. Unter trockenem Inertgas lagern. Nicht in der Nähe von Oxidationsmitteln, Wasser/Feuchtigkeit lagern.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Methylphthalic anhydride
Schmelzpunkt 114°C to 116°C
Menge 1 g
Empfindlichkeit Moisture sensitive
Löslichkeitsinformationen Reacts with water.
Formelmasse 162.14
Reinheit (%) 96%