missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)benzenboronsäure, 97 %, Thermo Scientific™

Catalog Number p-7048114
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
128312-11-8
C7H9BO2S
168.017
MFCD01319105
TYVPOLHSKGEXIH-UHFFFAOYSA-N
3-methylthio phenylboronic acid, 3-methylthio phenyl boronic acid, 3-meththio phenylboronic acid, 3-methylthiophenylboronic acid, 3-methylthio benzeneboronic acid, 3-methylsulfanyl phenylboronic acid, 3-boronothioanisole, 3-methylthiobenzeneboronic acid, 4-methylthiophenyl boronic acid, 3-methylthio phenyl-boronic acid
2734383
(3-methylsulfanylphenyl)Boronsäure
B(C1=CC(=CC=C1)SC)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
128312-11-8
C7H9BO2S
168.017
MFCD01319105
TYVPOLHSKGEXIH-UHFFFAOYSA-N
3-methylthio phenylboronic acid, 3-methylthio phenyl boronic acid, 3-meththio phenylboronic acid, 3-methylthiophenylboronic acid, 3-methylthio benzeneboronic acid, 3-methylsulfanyl phenylboronic acid, 3-boronothioanisole, 3-methylthiobenzeneboronic acid, 4-methylthiophenyl boronic acid, 3-methylthio phenyl-boronic acid
2734383
(3-methylsulfanylphenyl)Boronsäure
B(C1=CC(=CC=C1)SC)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(Methylthio)benzeneboronic acid
Schmelzpunkt 158°C to 161°C
Menge 250 mg
Beilstein 7020728
Formelmasse 168.02
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'