missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzenesulfonic acid, sodium salt, 97%

Product Code. 10656974 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10656974 2.5 kg Plastic drum 2.5kg
10246980 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656974 Supplier Thermo Scientific Acros Supplier No. 209120025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 127-68-4 | C6H4NNaO5S | 225.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-68-4
Molecular Formula C6H4NNaO5S
Molecular Weight (g/mol) 225.15
MDL Number MFCD00007490
InChI Key LJRGBERXYNQPJI-UHFFFAOYSA-M
Synonym sodium 3-nitrobenzenesulfonate, 3-nitrobenzenesulfonic acid sodium salt, sodium 3-nitrobenzenesulphonate, sodium m-nitrobenzenesulfonate, nitrol s, tiskan czech, ludigol, ludigol f,60, benzenesulfonic acid, 3-nitro-, sodium salt, unii-1f11sxj4c6
PubChem CID 31389
IUPAC Name sodium;3-nitrobenzenesulfonate
SMILES C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-].[Na+]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 350.0°C
Color Yellow
pH 6 to 10 (1% aq. soln.)
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic drum
Linear Formula O2NC6H4SO3Na
Quantity 2.5 kg
Beilstein 11, 68
Solubility Information Solubility in water: 200g/L water (20°C)
Formula Weight 225.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrobenzenesulfonic acid, sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.