missing translation for 'onlineSavingsMsg'
Learn More

3-O-Benzoyl-4,6-O-isopropyliden-D-glucal, 97 %, Thermo Scientific™

Catalog Number p-7040380
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
58871-20-8
C16H18O5
290.32
MFCD22988992
RADJUQJHMUFUJI-UHFFFAOYNA-N
3-o-benzoyl-4,6-o-isopropylidene-d-glucal, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl benzoate
92135621
[(4aR,8R,8aS)-2,2-Dimethyl-4,4a,8,8a-Tetrahydropyrano[3,2-d][1,3]Dioxin-8-yl]Benzoat
CC1(C)OCC2OC=CC(OC(=O)C3=CC=CC=C3)C2O1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
58871-20-8
C16H18O5
290.32
MFCD22988992
RADJUQJHMUFUJI-UHFFFAOYNA-N
3-o-benzoyl-4,6-o-isopropylidene-d-glucal, 4ar,8r,8as-2,2-dimethyl-4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-yl benzoate
92135621
[(4aR,8R,8aS)-2,2-Dimethyl-4,4a,8,8a-Tetrahydropyrano[3,2-d][1,3]Dioxin-8-yl]Benzoat
CC1(C)OCC2OC=CC(OC(=O)C3=CC=CC=C3)C2O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-O-Benzoyl-4,6-O-isopropylidene-D-glucal
Menge 250 mg
Formelmasse 290.31
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'