missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-2,3-dihydro-4H-1,4-benzoxazin-4-essigsäure, 98 %, Thermo Scientific™

Catalog Number p-7033209
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
26494-55-3
C10H9NO4
207.185
MFCD00269587
POGLODLVBYOXAO-UHFFFAOYSA-N
3-oxo-2,3-dihydro-4h-1,4-benzoxazin-4-yl acetic acid, 2-3-oxo-2h-benzo b 1,4 oxazin-4 3h-yl acetic acid, 2-3-oxo-3,4-dihydro-2h-1,4-benzoxazin-4-yl acetic acid, 3-oxo-2h-1,4-benzoxazin-4-yl acetic acid, acmc-1cmvy, 2-3-oxo-1,4-benzoxazin-4-yl acetic acid, 4-carboxymethyl-2h-1,4-benzoxazin-3 4h-one, 2-3-oxo-2h-benzo e 1,4-oxazin-4-yl acetic acid, 2,3-dihydro-3-oxo-4h-1,4-benzoxazine-4-acetic acid, 3-oxo-2,3-dihydro-4h-1,4-benzoxazine-4-acetic acid
2735705
2-(3-Oxo-H-1,4-Benzoxazin-4-yl)Ethansäure
C1C(=O)N(C2=CC=CC=C2O1)CC(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
26494-55-3
C10H9NO4
207.185
MFCD00269587
POGLODLVBYOXAO-UHFFFAOYSA-N
3-oxo-2,3-dihydro-4h-1,4-benzoxazin-4-yl acetic acid, 2-3-oxo-2h-benzo b 1,4 oxazin-4 3h-yl acetic acid, 2-3-oxo-3,4-dihydro-2h-1,4-benzoxazin-4-yl acetic acid, 3-oxo-2h-1,4-benzoxazin-4-yl acetic acid, acmc-1cmvy, 2-3-oxo-1,4-benzoxazin-4-yl acetic acid, 4-carboxymethyl-2h-1,4-benzoxazin-3 4h-one, 2-3-oxo-2h-benzo e 1,4-oxazin-4-yl acetic acid, 2,3-dihydro-3-oxo-4h-1,4-benzoxazine-4-acetic acid, 3-oxo-2,3-dihydro-4h-1,4-benzoxazine-4-acetic acid
2735705
2-(3-Oxo-H-1,4-Benzoxazin-4-yl)Ethansäure
C1C(=O)N(C2=CC=CC=C2O1)CC(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Oxo-2,3-dihydro-4H-1,4-benzoxazine-4-acetic acid
Menge 250 mg
Formelmasse 207.19
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'