missing translation for 'onlineSavingsMsg'
Learn More

3-(Pentafluorothio)zimtsäure, 97 %, Thermo Scientific™

Catalog Number p-7032370
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
773132-79-9
C9H7F5O2S
274.205
MFCD06205815
KMBHXZKXHIDSTE-SNAWJCMRSA-N
3-pentafluorosulfur cinnamic acid, 2e-3-3-pentafluoro-??-sulfanyl phenyl prop-2-enoic acid, 3-pentafluorothio cinnamic acid
66523449
(E)-3-[3-(Pentafluor-$l^{6}-sulfanyl)phenyl]prop-2-ensäure
C1=CC(=CC(=C1)S(F)(F)(F)(F)F)C=CC(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
773132-79-9
C9H7F5O2S
274.205
MFCD06205815
KMBHXZKXHIDSTE-SNAWJCMRSA-N
3-pentafluorosulfur cinnamic acid, 2e-3-3-pentafluoro-??-sulfanyl phenyl prop-2-enoic acid, 3-pentafluorothio cinnamic acid
66523449
(E)-3-[3-(Pentafluor-$l^{6}-sulfanyl)phenyl]prop-2-ensäure
C1=CC(=CC(=C1)S(F)(F)(F)(F)F)C=CC(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(Pentafluorothio)cinnamic acid
Menge 1 g
Formelmasse 274.21
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'