missing translation for 'onlineSavingsMsg'
Learn More

3-(Pentafluorthio)phenylessigsäure, 97 %

Catalog Number p-7032727
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1211578-68-5
C8H7F5O2S
262.194
MFCD16652414
MUXKXZMWGCIFAW-UHFFFAOYSA-N
3-pentafluorosulfur phenylacetic acid, 3-pentafluoro-??-sulfanyl phenyl acetic acid, 3-pentafluorothio phenylacetic acid
66523455
2-[3-(Pentafluor-$l^{6}-sulfanyl)phenyl]essigsäure
C1=CC(=CC(=C1)S(F)(F)(F)(F)F)CC(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1211578-68-5
C8H7F5O2S
262.194
MFCD16652414
MUXKXZMWGCIFAW-UHFFFAOYSA-N
3-pentafluorosulfur phenylacetic acid, 3-pentafluoro-??-sulfanyl phenyl acetic acid, 3-pentafluorothio phenylacetic acid
66523455
2-[3-(Pentafluor-$l^{6}-sulfanyl)phenyl]essigsäure
C1=CC(=CC(=C1)S(F)(F)(F)(F)F)CC(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(Pentafluorothio)phenylacetic acid
Menge 250 mg
Formelmasse 262.19
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'