missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-Propylacetat, 98 %, Thermo Scientific Chemicals

Catalog Number p-7025419
Click to view available options
Menge:
10 g
50 g
250 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
122-72-5
C11H14O2
178.231
MFCD00026216
JRJGKUTZNBZHNK-UHFFFAOYSA-N
hydrocinnamyl acetate, benzenepropanol, acetate, phenylpropyl acetate, 3-phenyl-1-propyl acetate, 3-acetoxypropyl benzene, 3-phenyl-1-propanol, acetate, 3-acetoxy-1-phenylpropane, 1-acetoxy-3-phenylpropane, benzenepropanol, 1-acetate, benzenepropyl acetate
31226
CHEBI:81257
3-Phenylpropyl-acetat
CC(=O)OCCCC1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
122-72-5
C11H14O2
178.231
MFCD00026216
JRJGKUTZNBZHNK-UHFFFAOYSA-N
hydrocinnamyl acetate, benzenepropanol, acetate, phenylpropyl acetate, 3-phenyl-1-propyl acetate, 3-acetoxypropyl benzene, 3-phenyl-1-propanol, acetate, 3-acetoxy-1-phenylpropane, 1-acetoxy-3-phenylpropane, benzenepropanol, 1-acetate, benzenepropyl acetate
31226
CHEBI:81257
3-Phenylpropyl-acetat
CC(=O)OCCCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Phenyl-1-propyl acetate
Dichte 1.01
Siedepunkt 252°C
Flammpunkt 130°C (266°F)
Brechungsindex 1.496
Menge 10 g
Formelmasse 178.23
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'