missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-Propylboronsäurepinacolester, 97 %, Thermo Scientific Chemicals

Catalog Number p-7030473
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
329685-40-7
C15H23BO2
246.16
MFCD09953505
HRZOKAQQKKQUME-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-3-phenylpropyl-1,3,2-dioxaborolane, 3-phenyl-1-propylboronic acid pinacol ester, 2-3-phenylpropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
15605871
4,4,5,5-tetramethyl-2-(3-phenylpropyl)-1,3,2-dioxaborolane
CC1(C)OB(CCCC2=CC=CC=C2)OC1(C)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
329685-40-7
C15H23BO2
246.16
MFCD09953505
HRZOKAQQKKQUME-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-3-phenylpropyl-1,3,2-dioxaborolane, 3-phenyl-1-propylboronic acid pinacol ester, 2-3-phenylpropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
15605871
4,4,5,5-tetramethyl-2-(3-phenylpropyl)-1,3,2-dioxaborolane
CC1(C)OB(CCCC2=CC=CC=C2)OC1(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Phenyl-1-propylboronic acid pinacol ester
Menge 1 g
Formelmasse 246.16
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'