missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluormethyl)pyridin-4-Carbonsäure, 95 %, Thermo Scientific Chemicals

Catalog Number p-7041659
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
590371-38-3
C7H4F3NO2
191.11
MFCD08234940
FBOSFEOQEZQFOC-UHFFFAOYSA-N
3-trifluoromethyl pyridine-4-carboxylic acid, 3-trifluoromethyl isonicotinic acid, 3-trifluoromethyl-isonicotinic acid, 3-trifluoromethyl-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3-trifluoromethyl, pubchem22216, ksc268s9d, 3-trifluoromethylisonicotinic acid, 3-trifluoromethyl isonicotinicacid
25210500
3-(Trifluormethyl)Pyridin-4-Carbonsäure
OC(=O)C1=C(C=NC=C1)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
590371-38-3
C7H4F3NO2
191.11
MFCD08234940
FBOSFEOQEZQFOC-UHFFFAOYSA-N
3-trifluoromethyl pyridine-4-carboxylic acid, 3-trifluoromethyl isonicotinic acid, 3-trifluoromethyl-isonicotinic acid, 3-trifluoromethyl-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 3-trifluoromethyl, pubchem22216, ksc268s9d, 3-trifluoromethylisonicotinic acid, 3-trifluoromethyl isonicotinicacid
25210500
3-(Trifluormethyl)Pyridin-4-Carbonsäure
OC(=O)C1=C(C=NC=C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-(Trifluoromethyl)pyridine-4-carboxylic acid
Menge 250 mg
Formelmasse 191.11
Reinheit (%) 95%

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'