missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinylcarbonylamino)Benzenboronsäure Pinacolester, 97 %, Thermo Scientific™

Catalog Number p-7034800
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
874290-95-6
C17H25BN2O3
316.208
MFCD08689514
YYWNYUFLSXVMRD-UHFFFAOYSA-N
n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine-1-carboxamide, 4-pyrrolidin-1-ylcarbonyl amino benzeneboronic acid, pinacol ester, n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine-1-carboxamide, 4-pyrrolidinylcarbonylamino phenylboronic acid, pinacol ester, 4-1-pyrrolidinylcarbonylamino benzeneboronic acid pinacol ester, 1-pyrrolidinecarboxamide, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl, 1-pyrrolidinecarboxamide,n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl
44119373
N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidin-1-carboxamid
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC(=O)N3CCCC3
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
874290-95-6
C17H25BN2O3
316.208
MFCD08689514
YYWNYUFLSXVMRD-UHFFFAOYSA-N
n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine-1-carboxamide, 4-pyrrolidin-1-ylcarbonyl amino benzeneboronic acid, pinacol ester, n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine-1-carboxamide, 4-pyrrolidinylcarbonylamino phenylboronic acid, pinacol ester, 4-1-pyrrolidinylcarbonylamino benzeneboronic acid pinacol ester, 1-pyrrolidinecarboxamide, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl, 1-pyrrolidinecarboxamide,n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl
44119373
N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidin-1-carboxamid
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC(=O)N3CCCC3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(1-Pyrrolidinylcarbonylamino)benzeneboronic acid pinacol ester
Schmelzpunkt 243°C to 245°C
Menge 250 mg
Formelmasse 316.21
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'