missing translation for 'onlineSavingsMsg'
Learn More

4-1BB Ligand/TNFSF9 Antibody (014), DyLight 680, Novus Biologicals™

Product Code. 18281258 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
0.1 mL
missing translation for 'unitSize'
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18281258 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18281258 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP290388FR

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

4-1BB Ligand/TNFSF9 Monoclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-1BB Ligand/TNFSF9
Anwendungen Flow Cytometry, ELISA, Immunofluorescence
Klassifikation Monoclonal
Klon 14
Konjugat DyLight 680
Verdünnung Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Zusammensetzung 50mM Sodium Borate
Gen-Alias 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9
Wirtsspezies Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse4-1BB Ligand/TNFSF9 (Arg 104-Glu 309; NP_033430.1).
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Apoptosis, Biologically Active Proteins
Primär oder sekundär Primary
Gen-ID (Entrez) 8744
Zielspezies Mouse
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.