missing translation for 'onlineSavingsMsg'
Learn More

4-(2,5-Dimethylphenylsulfonylamino)Benzoesäure, 96 %, Thermo Scientific™

Catalog Number p-7032100
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
126146-01-8
C15H15NO4S
305.35
MFCD02706676
MGFQTAXIYWVXQN-UHFFFAOYSA-N
4-2,5-dimethylbenzenesulfonamido benzoic acid, 4-2,5-dimethyl-benzenesulfonylamino-benzoic acid, 4-2,5-dimethylphenyl sulfonylamino benzoic acid, benzoic acid,4-2,5-dimethylphenyl sulfonyl amino, 4-2,5-dimethylphenylsulfonylamino benzoic acid, 4-2,5-dimethylphenyl sulfonamido benzoic acid, acmc-1c48m, 4-2,5-dimethylphenylsulfonamido benzoic acid, 4-2,5-dimethylphenyl sulfonyl amino benzoic acid, 4-2,5-dimethylbenzene-1-sulfonyl amino benzoic acid
978029
4-(2,5-dimethylbenzenesulfonamido)benzoic acid
CC1=CC(=C(C)C=C1)S(=O)(=O)NC1=CC=C(C=C1)C(O)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
126146-01-8
C15H15NO4S
305.35
MFCD02706676
MGFQTAXIYWVXQN-UHFFFAOYSA-N
4-2,5-dimethylbenzenesulfonamido benzoic acid, 4-2,5-dimethyl-benzenesulfonylamino-benzoic acid, 4-2,5-dimethylphenyl sulfonylamino benzoic acid, benzoic acid,4-2,5-dimethylphenyl sulfonyl amino, 4-2,5-dimethylphenylsulfonylamino benzoic acid, 4-2,5-dimethylphenyl sulfonamido benzoic acid, acmc-1c48m, 4-2,5-dimethylphenylsulfonamido benzoic acid, 4-2,5-dimethylphenyl sulfonyl amino benzoic acid, 4-2,5-dimethylbenzene-1-sulfonyl amino benzoic acid
978029
4-(2,5-dimethylbenzenesulfonamido)benzoic acid
CC1=CC(=C(C)C=C1)S(=O)(=O)NC1=CC=C(C=C1)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(2,5-Dimethylphenylsulfonylamino)benzoic acid
Schmelzpunkt 226°C to 227°C
Menge 250 mg
Formelmasse 305.35
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'