missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Carbamothioylhydrazinocarbonyl)benzenboronsäure, 98 %, Thermo Scientific™

Catalog Number p-7035378
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
957060-76-3
C8H10BN3O3S
239.056
MFCD09027244
NVXNXTLWXKMYHB-UHFFFAOYSA-N
2-4-boronobenzoyl hydrazinecarbothioamide, 4-2-carbamothioylhydrazinecarbonyl phenylboronic acid, 4-2-carbamothioylhydrazino carbonyl benzeneboronic acid, 4-carbamothioylaminocarbamoyl phenylboronic acid, 4-carbamothioylamino carbamoyl phenyl boronic acid, 4-2-carbamothioylhydrazinecarbonyl phenyl boronic acid, 4-2-carbamothioylhydrazinecarbonyl phenylboronic
44119176
[4-[(carbamothioylamino)carbamoyl]phenyl]boronsäure
B(C1=CC=C(C=C1)C(=O)NNC(=S)N)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
957060-76-3
C8H10BN3O3S
239.056
MFCD09027244
NVXNXTLWXKMYHB-UHFFFAOYSA-N
2-4-boronobenzoyl hydrazinecarbothioamide, 4-2-carbamothioylhydrazinecarbonyl phenylboronic acid, 4-2-carbamothioylhydrazino carbonyl benzeneboronic acid, 4-carbamothioylaminocarbamoyl phenylboronic acid, 4-carbamothioylamino carbamoyl phenyl boronic acid, 4-2-carbamothioylhydrazinecarbonyl phenyl boronic acid, 4-2-carbamothioylhydrazinecarbonyl phenylboronic
44119176
[4-[(carbamothioylamino)carbamoyl]phenyl]boronsäure
B(C1=CC=C(C=C1)C(=O)NNC(=S)N)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(2-Carbamothioylhydrazinocarbonyl)benzeneboronic acid
Schmelzpunkt 183°C to 184°C
Menge 250 mg
Formelmasse 239.06
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'