missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Cyano-3-Ethoxy-3-Oxo-1-Propen-1-yl)benzolboronsäurepinacolester, Thermo Scientific™

Catalog Number p-7033929
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1218790-51-2
C18H22BNO4
327.187
MFCD11113044
DNALFJBPPFISJQ-SDNWHVSQSA-N
ethyl 2e-2-cyano-3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl prop-2-enoate, 4-2-cyano-3-ethoxy-3-oxo-1-propen-1-yl benzeneboronic acid pinacol ester, 2-cyano-3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acrylic acid ethyl ester, amtb130, 2-cyano-3-4-4,4,5,5-tetramethyl-1,3,2 dioxa-borolan-2-yl-phenyl-acrylic acid ethyl ester, e-4-2-cyano-2-ethoxycarbonylvinyl phenyl boronic acid pinacol ester, e-ethyl 2-cyano-3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl acrylate, ethyl 2e-2-cyano-3-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl prop-2-enoate
46739750
Ethyl-(E)-2-cyan-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-enoat
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C=C(C#N)C(=O)OCC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1218790-51-2
C18H22BNO4
327.187
MFCD11113044
DNALFJBPPFISJQ-SDNWHVSQSA-N
ethyl 2e-2-cyano-3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl prop-2-enoate, 4-2-cyano-3-ethoxy-3-oxo-1-propen-1-yl benzeneboronic acid pinacol ester, 2-cyano-3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acrylic acid ethyl ester, amtb130, 2-cyano-3-4-4,4,5,5-tetramethyl-1,3,2 dioxa-borolan-2-yl-phenyl-acrylic acid ethyl ester, e-4-2-cyano-2-ethoxycarbonylvinyl phenyl boronic acid pinacol ester, e-ethyl 2-cyano-3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl acrylate, ethyl 2e-2-cyano-3-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl prop-2-enoate
46739750
Ethyl-(E)-2-cyan-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]prop-2-enoat
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C=C(C#N)C(=O)OCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(2-Cyano-3-ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid pinacol ester
Menge 1 g
Formelmasse 327.18
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'