missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Fluorbenzyl)-piperidin Hydrochlorid, 97 %, Thermo Scientific Chemicals

Catalog Number 15408556
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
193357-21-0
C12H17ClFN
229.72
MFCD08669673
XZRQMBCQGGXDKQ-UHFFFAOYSA-N
4-3-fluorobenzyl piperidine hydrochloride, 4-3-fluorophenyl methyl piperidine hydrochloride, 4-3-fluoro-benzyl-piperidine hydrochloride, 4-3-fluorobenzyl piperidine hcl
17039493
Cl.FC1=CC=CC(CC2CCNCC2)=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
193357-21-0
C12H17ClFN
229.72
MFCD08669673
XZRQMBCQGGXDKQ-UHFFFAOYSA-N
4-3-fluorobenzyl piperidine hydrochloride, 4-3-fluorophenyl methyl piperidine hydrochloride, 4-3-fluoro-benzyl-piperidine hydrochloride, 4-3-fluorobenzyl piperidine hcl
17039493
Cl.FC1=CC=CC(CC2CCNCC2)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(2-Fluorobenzyl)piperidine hydrochloride
Farbe Weiß
Menge 250 mg
IUPAC-Name 4-[(3-Fluorphenyl)Methyl]Piperidin;Hydrochlorid
Formelmasse 229.72
Reinheit (%) 97%
Physikalische Form Fest

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'