missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Fluorphenylsulfamoyl)benzenboronsäure, 97 %, Thermo Scientific™

Catalog Number p-7036374
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2096333-55-8
C12H11BFNO4S
295.09
MFCD20265311
LGYPQEPGDPRHPJ-UHFFFAOYSA-N
4-2-fluorophenylsulfamoyl benzeneboronic acid
73995911
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2096333-55-8
C12H11BFNO4S
295.09
MFCD20265311
LGYPQEPGDPRHPJ-UHFFFAOYSA-N
4-2-fluorophenylsulfamoyl benzeneboronic acid
73995911

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(2-Fluorophenylsulfamoyl)benzeneboronic acid
Menge 250 mg
Formelmasse 295.09
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'